Product Details
- SNP ID
-
rs4795403
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:39929469 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- TGCTTTATTTAGCGGGAGTCCCTGT[C/T]GTCATCTTTCTCCTTCTCTCTTGGC
- Phenotype
-
MIM: 610075
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LRRC3C
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs73985230] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LRRC3C
- Gene Name
- leucine rich repeat containing 3C
- Gene
- ORMDL3
- Gene Name
- ORMDL sphingolipid biosynthesis regulator 3
There are no transcripts associated with this gene.
View Full Product Details