Product Details

SNP ID
rs2973051
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:37813438 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAATGAAAATCCCTTTTATGAAATG[C/T]GTAACAAGCTGTAATACAGGGCCTA
Phenotype
MIM: 600837
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GDNF PubMed Links

Gene Details

Gene
GDNF
Gene Name
glial cell derived neurotrophic factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000514.3 3069 UTR 3 NP_000505.1
NM_001190468.1 3069 UTR 3 NP_001177397.1
NM_001190469.1 3069 UTR 3 NP_001177398.1
NM_001278098.1 3069 UTR 3 NP_001265027.1
NM_199231.2 3069 UTR 3 NP_954701.1
XM_011514030.2 3069 Intron XP_011512332.1
XM_017009337.1 3069 Intron XP_016864826.1

View Full Product Details