Product Details

SNP ID
rs8064778
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.17:78234900 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTCCCTGTGGTTCCAGAAGGTAC[C/G]CATGTATATTTGTTCTCACGTTGCT
Phenotype
MIM: 603352
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
BIRC5 PubMed Links

Gene Details

Gene
BIRC5
Gene Name
baculoviral IAP repeat containing 5
There are no transcripts associated with this gene.

Gene
TMEM235
Gene Name
transmembrane protein 235
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204210.1 Intron NP_001191139.1
NM_001204211.1 Intron NP_001191140.1
NM_001204212.1 Intron NP_001191141.1
XM_011524620.2 Intron XP_011522922.1
XM_011524624.2 Intron XP_011522926.1
XM_017024459.1 Intron XP_016879948.1
XM_017024460.1 Intron XP_016879949.1
XM_017024461.1 Intron XP_016879950.1

View Full Product Details