Product Details

SNP ID
rs7144448
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:35827865 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATCTTGAAGAATGAGTAGAATTTGG[G/T]TTGGGGTGGTGGGTATCCCTGCAGG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
BRMS1L PubMed Links

Gene Details

Gene
BRMS1L
Gene Name
breast cancer metastasis-suppressor 1-like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032352.3 Intron NP_115728.2
XM_005268128.1 Intron XP_005268185.1
XM_006720274.2 Intron XP_006720337.1
XM_006720275.2 Intron XP_006720338.1
XM_017021703.1 Intron XP_016877192.1
XM_017021704.1 Intron XP_016877193.1
XM_017021705.1 Intron XP_016877194.1
XM_017021706.1 Intron XP_016877195.1

View Full Product Details