Product Details

SNP ID
rs273641
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:51252410 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGAAAGAAGAAATCAGAATTTTGTA[C/G]TTAAATGGATCATTACATCTCCAGC
Phenotype
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
SIGLECL1 PubMed Links
Additional Information
For this assay, SNP(s) [rs78561654] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
SIGLECL1
Gene Name
SIGLEC family like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308437.1 Intron NP_001295366.1
NM_173635.2 Intron NP_775906.1
XM_005258799.4 Intron XP_005258856.1
XM_011526821.2 Intron XP_011525123.1
XM_017026647.1 Intron XP_016882136.1

View Full Product Details