Product Details

SNP ID
rs4949391
Assay Type
Functionally tested
NCBI dbSNP Submissions
56
Location
Chr.1:31360859 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAAAATGGGAATAATAACTCATGAA[G/T]TTTGACTCTACAGACTATTATTGTA
Phenotype
MIM: 134651
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FABP3 PubMed Links

Gene Details

Gene
FABP3
Gene Name
fatty acid binding protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320996.1 Intron NP_001307925.1
NM_004102.4 Intron NP_004093.1
XM_011541007.2 Intron XP_011539309.1
Gene
ZCCHC17
Gene Name
zinc finger CCHC-type containing 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282566.1 Intron NP_001269495.1
NM_001282567.1 Intron NP_001269496.1
NM_001282568.1 Intron NP_001269497.1
NM_001282569.1 Intron NP_001269498.1
NM_001282570.1 Intron NP_001269499.1
NM_001282571.1 Intron NP_001269500.1
NM_001282572.1 Intron NP_001269501.1
NM_001282573.1 Intron NP_001269502.1
NM_001282574.1 Intron NP_001269503.1
NM_016505.3 Intron NP_057589.2
XM_006710681.3 Intron XP_006710744.1
XM_011541567.2 Intron XP_011539869.1

View Full Product Details