Product Details

SNP ID
rs967467
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.15:100547844 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATTACACCAAAAGGTTTAGACTAA[A/G]AGTCACATAATGTAGCGGTGGAACC
Phenotype
MIM: 615276
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
CERS3 PubMed Links

Gene Details

Gene
CERS3
Gene Name
ceramide synthase 3
There are no transcripts associated with this gene.

Gene
PRKXP1
Gene Name
protein kinase, X-linked, pseudogene 1
There are no transcripts associated with this gene.

View Full Product Details