Product Details

SNP ID
rs1063178
Assay Type
Validated
NCBI dbSNP Submissions
82
Location
Chr.1:161751917 on Build GRCh38
Set Membership
HapMap JSNP Validated
Context Sequence [VIC/FAM]
TGAAATTATACCAGGCAATGGGATA[C/T]GAAGTGGATACCTCTAGTGCAATTT
Phenotype
MIM: 604835
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DUSP12 PubMed Links

Gene Details

Gene
DUSP12
Gene Name
dual specificity phosphatase 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007240.2 544 Silent Mutation TAC,TAT Y170Y NP_009171.1
XM_005244862.2 544 Silent Mutation TAC,TAT Y40Y XP_005244919.1

View Full Product Details