Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001080395.2 | Intron | NP_001073864.2 | |||
NM_004920.2 | Intron | NP_004911.2 | |||
XM_006722193.2 | Intron | XP_006722256.1 | |||
XM_006722194.2 | Intron | XP_006722257.1 | |||
XM_006722195.2 | Intron | XP_006722258.1 | |||
XM_006722196.2 | Intron | XP_006722259.1 | |||
XM_011525509.2 | Intron | XP_011523811.1 | |||
XM_011525510.2 | Intron | XP_011523812.1 | |||
XM_011525511.2 | Intron | XP_011523813.1 | |||
XM_017025421.1 | Intron | XP_016880910.1 | |||
XM_017025422.1 | Intron | XP_016880911.1 |