Product Details

SNP ID
rs12623408
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.2:232705241 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAATTCGCAGGGTCCAACTTCATAG[A/G]CTCTATTCACATTATTAGAGTAGTA
Phenotype
MIM: 612003
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
GIGYF2 PubMed Links
Additional Information
For this assay, SNP(s) [rs114285297] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
GIGYF2
Gene Name
GRB10 interacting GYF protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001103146.1 Intron NP_001096616.1
NM_001103147.1 Intron NP_001096617.1
NM_001103148.1 Intron NP_001096618.1
NM_015575.3 Intron NP_056390.2

View Full Product Details