Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001006939.3 | Intron | NP_001006940.3 | |||
XM_006717867.2 | Intron | XP_006717930.1 | |||
XM_011539822.1 | Intron | XP_011538124.1 | |||
XM_011539823.1 | Intron | XP_011538125.1 | |||
XM_011539826.2 | Intron | XP_011538128.1 | |||
XM_011539828.2 | Intron | XP_011538130.1 | |||
XM_011539829.2 | Intron | XP_011538131.1 | |||
XM_017016276.1 | Intron | XP_016871765.1 | |||
XM_017016277.1 | Intron | XP_016871766.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_020945.1 | Intron | NP_065996.1 | |||
XM_005270004.3 | Intron | XP_005270061.1 | |||
XM_011539986.2 | Intron | XP_011538288.2 | |||
XM_011539987.2 | Intron | XP_011538289.2 | |||
XM_011539988.2 | Intron | XP_011538290.1 | |||
XM_011539990.2 | Intron | XP_011538292.1 | |||
XM_011539991.2 | Intron | XP_011538293.1 | |||
XM_011539992.2 | Intron | XP_011538294.1 | |||
XM_017016463.1 | Intron | XP_016871952.1 | |||
XM_017016464.1 | Intron | XP_016871953.1 | |||
XM_017016465.1 | Intron | XP_016871954.1 |