Product Details

SNP ID
rs11961205
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:111102520 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATACAAGAGGACATATGCCAGTGA[A/G]TATATTAGAGTAAAACTTCATTCCC
Phenotype
MIM: 607550
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC16A10 PubMed Links

Gene Details

Gene
SLC16A10
Gene Name
solute carrier family 16 member 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018593.4 Intron NP_061063.2
XM_006715329.2 Intron XP_006715392.1
XM_011535422.2 Intron XP_011533724.1
XM_017010237.1 Intron XP_016865726.1

View Full Product Details