Product Details
- SNP ID
-
rs955462
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:6755827 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- GGAGAGGCCTGGCCTCAGGCTGCCA[C/G]CCTCAGGGAGGTAGATGCGGCTGTG
- Phenotype
-
MIM: 606717
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
XAF1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs3785790,rs78249496] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- XAF1
- Gene Name
- XIAP associated factor 1
There are no transcripts associated with this gene.
View Full Product Details