Product Details

SNP ID
rs2046615
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:219437563 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTTGACATTCATATGAGAATAGGA[A/G]GAGGACTGGGCTGCAGCAAATAACC
Phenotype
MIM: 615950
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SPEG PubMed Links

Gene Details

Gene
SPEG
Gene Name
SPEG complex locus
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001173476.1 Intron NP_001166947.1
NM_005876.4 Intron NP_005867.3
XM_005246237.2 Intron XP_005246294.1
XM_005246239.2 Intron XP_005246296.1
XM_005246240.2 Intron XP_005246297.1
XM_005246241.1 Intron XP_005246298.1
XM_005246242.4 Intron XP_005246299.1
XM_006712189.3 Intron XP_006712252.1
XM_006712193.3 Intron XP_006712256.1
XM_011510479.2 Intron XP_011508781.1
XM_011510483.2 Intron XP_011508785.2
XM_017003157.1 Intron XP_016858646.1
XM_017003158.1 Intron XP_016858647.1
XM_017003159.1 Intron XP_016858648.1
XM_017003160.1 Intron XP_016858649.1
XM_017003161.1 Intron XP_016858650.1
XM_017003162.1 Intron XP_016858651.1

View Full Product Details