Product Details

SNP ID
rs3168046
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:1275419 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATACCATCTAGAACGTTCCAGTCCC[A/G]CCTGACACCTTCATTCCCAATGGAG
Phenotype
MIM: 606277
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
TOLLIP PubMed Links

Gene Details

Gene
TOLLIP
Gene Name
toll interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318512.1 2609 UTR 3 NP_001305441.1
NM_001318514.1 2609 UTR 3 NP_001305443.1
NM_001318515.1 2609 UTR 3 NP_001305444.1
NM_001318516.1 2609 UTR 3 NP_001305445.1
NM_019009.3 2609 UTR 3 NP_061882.2
XM_017017931.1 2609 UTR 3 XP_016873420.1

View Full Product Details