Product Details

SNP ID
rs2001487
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:1856282 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAACCATTCACCAGCTACAGGCAAA[C/G]TCCATGCCCCTGGGTGGCCATGCCA
Phenotype
MIM: 153432
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LSP1 PubMed Links

Gene Details

Gene
LSP1
Gene Name
lymphocyte-specific protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013253.1 Intron NP_001013271.1
NM_001013254.1 Intron NP_001013272.1
NM_001013255.1 Intron NP_001013273.1
NM_001242932.1 Intron NP_001229861.1
NM_001289005.1 Intron NP_001275934.1
NM_002339.2 Intron NP_002330.1
Gene
MIR4298
Gene Name
microRNA 4298
There are no transcripts associated with this gene.

View Full Product Details