Product Details
- SNP ID
-
hCV1608295
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:15304198 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CATGGTTAGGTCAGCATGTAGCTTA[C/T]GCAACTCCAAGTACATCACATTGTA
- Phenotype
-
MIM: 612683
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TEKT3
PubMed Links
Gene Details
- Gene
- TEKT3
- Gene Name
- tektin 3
There are no transcripts associated with this gene.
View Full Product Details