Product Details

SNP ID
rs35586271
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:92844962 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGAATCCAAATATACAAAATTAAT[A/C]CCATATAAGAGACAAATAAATGAAA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ANKRD19P PubMed Links

Gene Details

Gene
ANKRD19P
Gene Name
ankyrin repeat domain 19, pseudogene
There are no transcripts associated with this gene.

Gene
ZNF484
Gene Name
zinc finger protein 484
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007101.2 4108 Intron NP_001007102.1
NM_001261458.1 4108 Intron NP_001248387.1
NM_001261459.1 4108 Intron NP_001248388.1
NM_001261460.1 4108 Intron NP_001248389.1
NM_031486.2 4108 Intron NP_113674.1
XM_006717299.3 4108 UTR 3 XP_006717362.1
XM_011519073.2 4108 UTR 3 XP_011517375.1
XM_011519074.2 4108 UTR 3 XP_011517376.1
XM_011519075.2 4108 UTR 3 XP_011517377.1
XM_017015178.1 4108 UTR 3 XP_016870667.1
XM_017015179.1 4108 UTR 3 XP_016870668.1
XM_017015180.1 4108 UTR 3 XP_016870669.1
XM_017015181.1 4108 UTR 3 XP_016870670.1

View Full Product Details