Product Details

SNP ID
rs10817477
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.9:113313517 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GGAGCCAGAAAGTTAATAACTGGTC[C/T]TGAAGATACAAACAATGTCATGGTC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
WDR31 PubMed Links

Gene Details

Gene
WDR31
Gene Name
WD repeat domain 31
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001006615.2 4638 UTR 3 NP_001006616.1
NM_001012361.3 4638 UTR 3 NP_001012361.1
NM_145241.4 4638 UTR 3 NP_660284.1
XM_011518194.2 4638 Intron XP_011516496.1

View Full Product Details