Product Details

SNP ID
rs10893329
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:125112127 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGAACATCTCAATCCCCAACCCACT[A/G]GCTCGAGTGGGGCGCAAGATGTGGT
Phenotype
MIM: 613066
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
KRT18P59 PubMed Links

Gene Details

Gene
KRT18P59
Gene Name
keratin 18 pseudogene 59
There are no transcripts associated with this gene.

Gene
PKNOX2
Gene Name
PBX/knotted 1 homeobox 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022062.2 131 Intron NP_071345.2
XM_005271642.2 131 Intron XP_005271699.1
XM_005271643.2 131 Intron XP_005271700.1
XM_006718894.2 131 Intron XP_006718957.1
XM_011542944.2 131 Intron XP_011541246.1
XM_011542945.2 131 Intron XP_011541247.1
XM_011542946.1 131 Intron XP_011541248.1
XM_011542947.2 131 Intron XP_011541249.1
XM_017018110.1 131 UTR 5 XP_016873599.1
XM_017018111.1 131 Intron XP_016873600.1
XM_017018112.1 131 Intron XP_016873601.1
Gene
TMEM218
Gene Name
transmembrane protein 218
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080546.2 131 Intron NP_001074015.1
NM_001258238.1 131 Intron NP_001245167.1
NM_001258239.1 131 Intron NP_001245168.1
NM_001258240.1 131 Intron NP_001245169.1
NM_001258241.1 131 Intron NP_001245170.1
NM_001258242.1 131 Intron NP_001245171.1
NM_001258243.1 131 Intron NP_001245172.1
NM_001258244.1 131 Intron NP_001245173.1
NM_001258245.1 131 Intron NP_001245174.1
NM_001258246.1 131 Intron NP_001245175.1
NM_001258247.1 131 Intron NP_001245176.1
XM_006718779.3 131 Intron XP_006718842.1
XM_006718780.3 131 Intron XP_006718843.1
XM_006718781.3 131 Intron XP_006718844.1
XM_006718783.3 131 Intron XP_006718846.1
XM_017017322.1 131 Intron XP_016872811.1
XM_017017323.1 131 Intron XP_016872812.1
XM_017017324.1 131 Intron XP_016872813.1

View Full Product Details