Product Details

SNP ID
rs2305778
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.19:13933474 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CAGCTTGCTGGAAGGAGAGAGGGTC[A/G]GTGTTAGGTGGGGCACTTGGAGTGG
Phenotype
MIM: 610055
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CC2D1A PubMed Links

Gene Details

Gene
CC2D1A
Gene Name
coiled-coil and C2 domain containing 1A
There are no transcripts associated with this gene.

Gene
PODNL1
Gene Name
podocan like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146254.1 Intron NP_001139726.1
NM_001146255.1 Intron NP_001139727.1
NM_024825.3 Intron NP_079101.3
XM_006722901.1 Intron XP_006722964.1
XM_006722902.1 Intron XP_006722965.1
XM_006722903.1 Intron XP_006722966.1
XM_011528308.1 Intron XP_011526610.1
XM_011528309.2 Intron XP_011526611.1
XM_011528310.2 Intron XP_011526612.1
XM_011528311.1 Intron XP_011526613.1
XM_011528313.1 Intron XP_011526615.1
XM_011528314.1 Intron XP_011526616.1
XM_011528316.1 Intron XP_011526618.1
XM_017027313.1 Intron XP_016882802.1

View Full Product Details