Product Details
- SNP ID
-
rs1048690
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:35125112 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCCACGGTCAGCAGCTCACAGGCGG[A/G]CCATCACCCCAAGTAGGGCCAGGAG
- Phenotype
-
MIM: 604996
MIM: 608303
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
FXYD3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs370592226] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FXYD3
- Gene Name
- FXYD domain containing ion transport regulator 3
- Gene
- LGI4
- Gene Name
- leucine rich repeat LGI family member 4
- Gene
- MIR6887
- Gene Name
- microRNA 6887
There are no transcripts associated with this gene.
View Full Product Details