Product Details

SNP ID
rs6764352
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:149039038 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACTAAGATTCTGAAAAAATTTACA[A/G]AACTACTTACTGGATCCATTAAAAA
Phenotype
MIM: 603257
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HLTF PubMed Links

Gene Details

Gene
HLTF
Gene Name
helicase like transcription factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318934.1 Intron NP_001305863.1
NM_001318935.1 Intron NP_001305864.1
NM_003071.3 Intron NP_003062.2
NM_139048.2 Intron NP_620636.1
XM_011513091.1 Intron XP_011511393.1
XM_011513092.1 Intron XP_011511394.1
XM_011513093.1 Intron XP_011511395.1
XM_017007078.1 Intron XP_016862567.1
XM_017007079.1 Intron XP_016862568.1

View Full Product Details