Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001318934.1 | Intron | NP_001305863.1 | |||
NM_001318935.1 | Intron | NP_001305864.1 | |||
NM_003071.3 | Intron | NP_003062.2 | |||
NM_139048.2 | Intron | NP_620636.1 | |||
XM_011513091.1 | Intron | XP_011511393.1 | |||
XM_011513092.1 | Intron | XP_011511394.1 | |||
XM_011513093.1 | Intron | XP_011511395.1 | |||
XM_017007078.1 | Intron | XP_016862567.1 | |||
XM_017007079.1 | Intron | XP_016862568.1 |