Product Details

SNP ID
rs34572934
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:5783504 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCACCTTGAGCTCGCTGAGGCCC[A/G]GTGGGTACAGGGGCACCCCAGGTCG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CATSPERD PubMed Links

Gene Details

Gene
CATSPERD
Gene Name
cation channel sperm associated auxiliary subunit delta
There are no transcripts associated with this gene.

Gene
DUS3L
Gene Name
dihydrouridine synthase 3 like
There are no transcripts associated with this gene.

Gene
PRR22
Gene Name
proline rich 22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134316.1 848 Missense Mutation CCG,CTG P248L NP_001127788.1

View Full Product Details