Product Details

SNP ID
rs2125341
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:172836523 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTGCACCTTGGAGGATGAGCCATGT[A/G]TGCCCAGCAAACCTGCACTCTTTAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ERGIC1 PubMed Links

Gene Details

Gene
ERGIC1
Gene Name
endoplasmic reticulum-golgi intermediate compartment 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031711.2 Intron NP_001026881.1
XM_006714892.1 Intron XP_006714955.1
XM_011534597.1 Intron XP_011532899.1
XM_011534598.2 Intron XP_011532900.1
XM_017009654.1 Intron XP_016865143.1
XM_017009655.1 Intron XP_016865144.1

View Full Product Details