Product Details

SNP ID
rs10899922
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:43846516 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CGGGCAGCTGGAAGGGCATCCTGAA[A/G]AGACGCCTGCCTGCCGCCGTTCTCC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LINC00619 PubMed Links

Gene Details

Gene
LINC00619
Gene Name
long intergenic non-protein coding RNA 619
There are no transcripts associated with this gene.

View Full Product Details