Product Details

SNP ID
rs3768025
Assay Type
Functionally tested
NCBI dbSNP Submissions
39
Location
Chr.1:42661062 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
AGAGAATGGTGATGGGGAAGTTCTT[A/G]AAAGTGGCTGGGTACGACTGACTGT
Phenotype
MIM: 606095
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC30 PubMed Links

Gene Details

Gene
CCDC30
Gene Name
coiled-coil domain containing 30
There are no transcripts associated with this gene.

Gene
PPIH
Gene Name
peptidylprolyl isomerase H
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006347.3 Intron NP_006338.1
XM_005270362.1 Intron XP_005270419.1
XM_005270366.3 Intron XP_005270423.1
XM_011540508.1 Intron XP_011538810.1
XM_017000057.1 Intron XP_016855546.1
XM_017000058.1 Intron XP_016855547.1
XM_017000059.1 Intron XP_016855548.1

View Full Product Details