Product Details

SNP ID
rs12589873
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:24011912 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GATGCACCATCTGCCTGATTTTTTC[C/T]GAATTACAAACGCAGGAGAATGCCA
Phenotype
MIM: 615195 MIM: 615196
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DHRS4L1 PubMed Links

Gene Details

Gene
DHRS4L1
Gene Name
dehydrogenase/reductase 4 like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277864.1 Intron NP_001264793.1
XM_006720246.3 Intron XP_006720309.1
Gene
DHRS4L2
Gene Name
dehydrogenase/reductase 4 like 2
There are no transcripts associated with this gene.

View Full Product Details