Product Details

SNP ID
rs8074016
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.17:58763226 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGAGCTGTGTTCTGTGAAGTCATTA[A/G]GGAACCCAGGCTGATGGAGACTCTG
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
PPM1E PubMed Links

Gene Details

Gene
PPM1E
Gene Name
protein phosphatase, Mg2+/Mn2+ dependent 1E
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014906.4 Intron NP_055721.3
XM_005257169.4 Intron XP_005257226.1
XM_011524534.2 Intron XP_011522836.1
XM_017024359.1 Intron XP_016879848.1

View Full Product Details