Product Details

SNP ID
rs11889208
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:205689798 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGTGTGCTCAGCAGGGAACACCG[C/T]GTTTAAGGCGCTCTCCCGAATGTGT
Phenotype
MIM: 602070
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NRP2 PubMed Links

Gene Details

Gene
NRP2
Gene Name
neuropilin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003872.2 Intron NP_003863.2
NM_018534.3 Intron NP_061004.3
NM_201264.1 Intron NP_957716.1
NM_201266.1 Intron NP_957718.1
NM_201267.1 Intron NP_957719.1
NM_201279.1 Intron NP_958436.1
XM_005246933.3 Intron XP_005246990.2
XM_005246934.3 Intron XP_005246991.2
XM_017005185.1 Intron XP_016860674.1
XM_017005186.1 Intron XP_016860675.1
XM_017005187.1 Intron XP_016860676.1
XM_017005188.1 Intron XP_016860677.1

View Full Product Details