Product Details
- SNP ID
-
rs2676395
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:33485791 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGGATTTGGTTAACCAGAAAAAATG[C/T]CCCACCTCTCCTGGAACACTGATGG
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MAK16
PubMed Links
Gene Details
- Gene
- MAK16
- Gene Name
- MAK16 homolog
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_032509.3 |
|
Intron |
|
|
NP_115898.2 |
View Full Product Details