Product Details

SNP ID
rs11800843
Assay Type
Functionally tested
NCBI dbSNP Submissions
38
Location
Chr.1:169369793 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTAAACATTTAATGTTAATGCAAA[A/G]TTTTTAACAGCTATATTCCCTATAC
Phenotype
MIM: 608692 MIM: 613465
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BLZF1 PubMed Links

Gene Details

Gene
BLZF1
Gene Name
basic leucine zipper nuclear factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320972.1 Intron NP_001307901.1
NM_001320973.1 Intron NP_001307902.1
NM_003666.3 Intron NP_003657.1
XM_005245561.4 Intron XP_005245618.1
XM_011510092.2 Intron XP_011508394.1
XM_017002645.1 Intron XP_016858134.1
Gene
NME7
Gene Name
NME/NM23 family member 7
There are no transcripts associated with this gene.

View Full Product Details