Product Details

SNP ID
rs10906083
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:12137271 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCAAATTTTTGCAATCTTGGGTTGT[A/G]CTGTGTTTAAAAATTACTAAAATTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR548AK PubMed Links

Gene Details

Gene
MIR548AK
Gene Name
microRNA 548ak
There are no transcripts associated with this gene.

Gene
SEC61A2
Gene Name
Sec61 translocon alpha 2 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142627.2 Intron NP_001136099.1
NM_001142628.1 Intron NP_001136100.1
NM_018144.3 Intron NP_060614.2

View Full Product Details