Product Details

SNP ID
rs2231441
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:118304825 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCTTTTTCATCCTAGCATTGGGA[A/G]CAATGGCCCCAGGGTCCTTATCTCT
Phenotype
MIM: 186830
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CD3E PubMed Links

Gene Details

Gene
CD3E
Gene Name
CD3e molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000733.3 Intron NP_000724.1

View Full Product Details