Product Details

SNP ID
rs12676593
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:58561379 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTGGACATCCAGATTCAAGAAGCCA[A/G]ATGGACTCCAGAGTAGAACAATCCG
Phenotype
MIM: 602217
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SDCBP PubMed Links

Gene Details

Gene
SDCBP
Gene Name
syndecan binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007067.1 1091 Intron NP_001007068.1
NM_001007068.1 1091 Intron NP_001007069.1
NM_001007069.1 1091 Intron NP_001007070.1
NM_001007070.1 1091 Intron NP_001007071.1
NM_005625.3 1091 Intron NP_005616.2
XM_011517571.2 1091 UTR 5 XP_011515873.2
XM_017013727.1 1091 UTR 5 XP_016869216.1
XM_017013728.1 1091 UTR 5 XP_016869217.1

View Full Product Details