Product Details
- SNP ID
-
rs11914833
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:180616931 on Build GRCh38
- Set Membership
-
Validated
- Context Sequence [VIC/FAM]
- ATAACTTTTCTTTAACATTATTTGC[C/T]AAATGTTCTATAACATCTAATGTAT
- Phenotype
-
MIM: 613798
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CCDC39
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs79386936] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CCDC39
- Gene Name
- coiled-coil domain containing 39
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_181426.1 |
2416 |
Silent Mutation |
TTA,TTG |
L767L |
NP_852091.1 |
- Gene
- TTC14
- Gene Name
- tetratricopeptide repeat domain 14
View Full Product Details