Product Details

SNP ID
rs10124070
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.9:98289597 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
GTCTACGGGGATGTCTTTTAGCAGA[A/G]TGCCTCCAATTCACTTTGGTCAGAA
Phenotype
MIM: 607340
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GABBR2 PubMed Links

Gene Details

Gene
GABBR2
Gene Name
gamma-aminobutyric acid type B receptor subunit 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005458.7 4273 UTR 3 NP_005449.5
XM_005252316.4 4273 Intron XP_005252373.1
XM_017015331.1 4273 Intron XP_016870820.1
XM_017015332.1 4273 Intron XP_016870821.1

View Full Product Details