Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001184940.1 | Intron | NP_001171869.1 | |||
NM_001184941.1 | Intron | NP_001171870.1 | |||
NM_001184942.1 | Intron | NP_001171871.1 | |||
NM_001184943.1 | Intron | NP_001171872.1 | |||
NM_001184945.1 | Intron | NP_001171874.1 | |||
NM_147202.1 | Intron | NP_671735.1 | |||
XM_005251402.4 | Intron | XP_005251459.1 | |||
XM_011517797.2 | Intron | XP_011516099.1 | |||
XM_011517798.2 | Intron | XP_011516100.1 | |||
XM_011517799.2 | Intron | XP_011516101.1 |