Product Details

SNP ID
rs12266534
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:129471492 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGGGGATATCATGTTCAGGTGTCT[C/T]TCCTGCTTGTGTGGGGTAAGAGCCA
Phenotype
MIM: 156569
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MGMT PubMed Links

Gene Details

Gene
MGMT
Gene Name
O-6-methylguanine-DNA methyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002412.4 Intron NP_002403.2
XM_005252682.2 Intron XP_005252739.1
XM_017016275.1 Intron XP_016871764.1

View Full Product Details