Product Details

SNP ID
rs4749125
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:26438707 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCGGATAGCGCCACACGTCTGCGC[C/G]CTGCGTGGGAAGGGCAGGGCTGACA
Phenotype
MIM: 609036
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
APBB1IP PubMed Links

Gene Details

Gene
APBB1IP
Gene Name
amyloid beta precursor protein binding family B member 1 interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019043.3 175 UTR 5 NP_061916.3
XM_006717451.2 175 Intron XP_006717514.1
XM_011519514.2 175 UTR 5 XP_011517816.1

View Full Product Details