Product Details

SNP ID
rs2947594
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:122697936 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCGATGGACCGTTCCGGTTTCTTTG[C/T]GGGGAGGAATGATTTGCGATCATTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C10orf120 PubMed Links

Gene Details

Gene
C10orf120
Gene Name
chromosome 10 open reading frame 120
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010912.2 837 Missense Mutation ACA,GCA T269A NP_001010912.1

View Full Product Details