Product Details
- SNP ID
-
rs10888486
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
55
- Location
-
Chr.1:152409586 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- TTCTTCCAGTACTGGACATTGGAGT[C/T]GGGGAAGTCATGGCTTGGTGCTTCT
- Phenotype
-
MIM: 611312
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CRNN
PubMed Links
Gene Details
- Gene
- CRNN
- Gene Name
- cornulin
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_016190.2 |
1570 |
UTR 3 |
|
|
NP_057274.1 |
View Full Product Details