Product Details

SNP ID
rs17567355
Assay Type
Validated
NCBI dbSNP Submissions
27
Location
Chr.1:109130398 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GCTTCACAGAAGGAGTAAGAGTCAC[A/G]GAGGGCTTCAAAGAAGTACTAAAAT
Phenotype
MIM: 611298
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KIAA1324 PubMed Links

Gene Details

Gene
KIAA1324
Gene Name
KIAA1324
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001267048.1 Intron NP_001253977.1
NM_001284352.1 Intron NP_001271281.1
NM_001284353.1 Intron NP_001271282.1
NM_020775.4 Intron NP_065826.2
XM_011541825.1 Intron XP_011540127.1
XM_011541826.2 Intron XP_011540128.1
XM_011541827.1 Intron XP_011540129.1
XM_017001883.1 Intron XP_016857372.1

View Full Product Details