Product Details
- SNP ID
-
rs667524
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
42
- Location
-
Chr.1:109114324 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCCAGGGAACGGGACCGGAGCTTCA[C/T]GCCTGCAAAGAGGTACTGCCGCCCC
- Phenotype
-
MIM: 611298
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
C1orf194
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs586530] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- C1orf194
- Gene Name
- chromosome 1 open reading frame 194
- Gene
- KIAA1324
- Gene Name
- KIAA1324
View Full Product Details