Product Details

SNP ID
rs1905055
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.11:56346040 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TTTCTAGTTTGCACAGTTACCAAGA[C/T]GAATCATGTGGTAAAACACAATCAC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR8K1 PubMed Links

Gene Details

Gene
OR8K1
Gene Name
olfactory receptor family 8 subfamily K member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002907.1 2 Missense Mutation ACG,ATG T1M NP_001002907.1

View Full Product Details