Product Details

SNP ID
rs35606247
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:13283855 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCTCCCCCTGCTTTCTCTGCAGC[A/G]TTGCTCTGACCCAGGCCCCCTGTGG
Phenotype
MIM: 602550
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARNTL PubMed Links

Gene Details

Gene
ARNTL
Gene Name
aryl hydrocarbon receptor nuclear translocator like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001030272.2 Intron NP_001025443.1
NM_001030273.2 Intron NP_001025444.1
NM_001178.5 Intron NP_001169.3
NM_001297719.1 Intron NP_001284648.1
NM_001297722.1 Intron NP_001284651.1
NM_001297724.1 Intron NP_001284653.1
XM_006718233.3 Intron XP_006718296.1
XM_006718234.1 Intron XP_006718297.1
XM_011520105.2 Intron XP_011518407.2
XM_011520107.2 Intron XP_011518409.2
XM_011520108.1 Intron XP_011518410.1
XM_011520109.1 Intron XP_011518411.1
XM_011520110.1 Intron XP_011518412.1
XM_011520111.1 Intron XP_011518413.1
XM_011520112.2 Intron XP_011518414.1
XM_011520113.1 Intron XP_011518415.1
XM_017017738.1 Intron XP_016873227.1
XM_017017739.1 Intron XP_016873228.1
XM_017017740.1 Intron XP_016873229.1
XM_017017741.1 Intron XP_016873230.1
XM_017017742.1 Intron XP_016873231.1
XM_017017743.1 Intron XP_016873232.1
XM_017017744.1 Intron XP_016873233.1
XM_017017745.1 Intron XP_016873234.1
XM_017017746.1 Intron XP_016873235.1
XM_017017747.1 Intron XP_016873236.1
XM_017017748.1 Intron XP_016873237.1
Gene
LOC105376560
Gene Name
uncharacterized LOC105376560
There are no transcripts associated with this gene.

View Full Product Details