Product Details

SNP ID
rs12257237
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:6393195 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGCTTTTTTTGTGAAGGGCCTGAGA[C/G]TGAATATTTTTCACCTTCACTTTTG
Phenotype
MIM: 600448
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PRKCQ PubMed Links

Gene Details

Gene
PRKCQ
Gene Name
protein kinase C theta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242413.2 3382 Intron NP_001229342.1
NM_001282644.1 3382 Intron NP_001269573.1
NM_001282645.1 3382 Intron NP_001269574.1
NM_001323265.1 3382 Intron NP_001310194.1
NM_001323266.1 3382 Intron NP_001310195.1
NM_001323267.1 3382 Intron NP_001310196.1
NM_006257.4 3382 Intron NP_006248.1
XM_005252496.4 3382 Intron XP_005252553.1
XM_005252497.4 3382 UTR 3 XP_005252554.1
XM_006717465.3 3382 Intron XP_006717528.1
XM_017016410.1 3382 Intron XP_016871899.1
XM_017016411.1 3382 Intron XP_016871900.1

View Full Product Details