Product Details

SNP ID
rs36004176
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:178539186 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGATATTTCTCAAACTAACACAA[A/T]CTTCCCTTTGATCATCTCTCCCTCT
Phenotype
MIM: 605214
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
KCNMB2 PubMed Links

Gene Details

Gene
KCNMB2
Gene Name
potassium calcium-activated channel subfamily M regulatory beta subunit 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278911.1 Intron NP_001265840.1
NM_005832.4 Intron NP_005823.1
NM_181361.2 Intron NP_852006.1
XM_011512325.2 Intron XP_011510627.1
Gene
KCNMB2-AS1
Gene Name
KCNMB2 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details