Product Details

SNP ID
rs9916085
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:81313677 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CACCTCCCTCTAATCCCTGCACCCC[A/G]GCCCCACAGCCCCTTCATCCTCACC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LINC00482 PubMed Links

Gene Details

Gene
LINC00482
Gene Name
long intergenic non-protein coding RNA 482
There are no transcripts associated with this gene.

Gene
TMEM105
Gene Name
transmembrane protein 105
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178520.3 914 Missense Mutation CGG,TGG R122W NP_848615.1
XM_017024522.1 914 Missense Mutation CGG,TGG R100W XP_016880011.1

View Full Product Details